Anti-p-Stat3 Rabbit Polyclonal Antibody [clone: S727]

Lieferant: Bioworld Technology
BS4180
BWRLBS4180EA 284 EUR
BWRLBS4180
Anti-p-Stat3 Rabbit Polyclonal Antibody [clone: S727]
Antikörper
Synthetic phosphopeptide derived from human Stat3 around the phosphorylation site of Serine 727.

p-Stat3 (S727) pAb detects endogenous levels of Stat3 protein only when phosphorylated at Ser727.

Type: Primary
Antigen: p-Stat3
Clonality: Polyclonal
Clone: S727
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR