Anti-p-Stat3 Rabbit Polyclonal Antibody [clone: Y705]

Lieferant: Bioworld Technology
BS4181
BWRLBS4181EA 284 EUR
BWRLBS4181
Anti-p-Stat3 Rabbit Polyclonal Antibody [clone: Y705]
Antikörper
Synthetic phosphopeptide derived from human Stat3 around the phosphorylation site of Tyrosine 705.

p-Stat3 (Y705) pAb detects endogenous levels of Stat3 protein only when phosphorylated at Tyr705

Type: Primary
Antigen: p-Stat3
Clonality: Polyclonal
Clone: Y705
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR