Anti-p-Synaptotagmin 1/2 Rabbit Polyclonal Antibody [clone: T202/199]

Lieferant: Bioworld Technology
BS4191
BWRLBS4191EA 284 EUR
BWRLBS4191
Anti-p-Synaptotagmin 1/2 Rabbit Polyclonal Antibody [clone: T202/199]
Antikörper
Synthetic phosphopeptide derived from human Synaptotagmin 1 around the phosphorylation site of Threonine 202.

p-Synaptotagmin 1/2 (T202/199) pAb detects endogenous levels of Synaptotagmin 1/2 protein only when phosphorylated at SerT202/199.

Type: Primary
Antigen: p-Synaptotagmin 1/2
Clonality: Polyclonal
Clone: T202/199
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR