Anti-p-Ku-80 Rabbit Polyclonal Antibody [clone: T714]

Lieferant: Bioworld Technology
BS4240
BWRLBS4240EA 284 EUR
BWRLBS4240
Anti-p-Ku-80 Rabbit Polyclonal Antibody [clone: T714]
Antikörper
Synthetic phosphopeptide derived from human Ku-80 around the phosphorylation site of Threonine 714.

p-Ku-80 (T714) pAb detects endogenous levels of Ku-80 protein only when phosphorylated at Thr714

Type: Primary
Antigen: p-Ku-80
Clonality: Polyclonal
Clone: T714
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR