Anti-p-Shc Rabbit Polyclonal Antibody [clone: Y427]

Lieferant: Bioworld Technology
BS4251
BWRLBS4251EA 284 EUR
BWRLBS4251
Anti-p-Shc Rabbit Polyclonal Antibody [clone: Y427]
Antikörper
Synthetic phosphopeptide derived from human Shc around the phosphorylation site of Tyrosine 427.

p-Shc (Y427) pAb detects endogenous levels of Shc protein only when phosphorylated at Tyr427.

Type: Primary
Antigen: p-Shc
Clonality: Polyclonal
Clone: Y427
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR