Anti-p-MITF Rabbit Polyclonal Antibody [clone: S180]

Lieferant: Bioworld Technology
BS4328
BWRLBS4328EA 284 EUR
BWRLBS4328
Anti-p-MITF Rabbit Polyclonal Antibody [clone: S180]
Antikörper
Synthetic phosphopeptide derived from human MITF around the phosphorylation site of Serine 180.

p-MITF (S180) pAb detects endogenous levels of MITF protein only when phosphorylated at Ser180

Type: Primary
Antigen: p-MITF
Clonality: Polyclonal
Clone: S180
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR