Anti-p-PPARgamma Rabbit Polyclonal Antibody [clone: S112]

Lieferant: Bioworld Technology
BS4444
BWRLBS4444EA 284 EUR
BWRLBS4444
Anti-p-PPARgamma Rabbit Polyclonal Antibody [clone: S112]
Antikörper
Synthetic phosphopeptide derived from human PPARgamma around the phosphorylation site of Serine 112.

p-PPAR? (S112) pAb detects endogenous levels of PPAR? protein only when phosphorylated at Ser112.

Type: Primary
Antigen: p-PPARgamma
Clonality: Polyclonal
Clone: S112
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR