Anti-p-FAK Rabbit Polyclonal Antibody [clone: Y397]
Lieferant: Bioworld Technology
BS4617
BWRLBS4617EA 284 EUR
BWRLBS4617
Anti-p-FAK Rabbit Polyclonal Antibody [clone: Y397]
Antikörper
Synthetic phosphopeptide derived from human FAK around the phosphorylation site of Serine 397.
p-FAK (Y397) pAb detects endogenous levels of FAK protein only when phosphorylated at Tyr397.
Type: Primary
Antigen: p-FAK
Clonality: Polyclonal
Clone: Y397
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat