Anti-p-APC Rabbit Polyclonal Antibody [clone: S2054]

Lieferant: Bioworld Technology
BS4649
BWRLBS4649EA 284 EUR
BWRLBS4649
Anti-p-APC Rabbit Polyclonal Antibody [clone: S2054]
Antikörper
Synthetic phosphopeptide derived from human APC around the phosphorylation site of Serine 2054.

p-APC (S2054) pAb detects endogenous levels of APC protein when phosphorylated at Ser2054.

Type: Primary
Antigen: p-APC
Clonality: Polyclonal
Clone: S2054
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR