Anti-p-ATF2 Rabbit Polyclonal Antibody [clone: S498]
Lieferant: Bioworld Technology
BS4654
BWRLBS4654EA 284 EUR
BWRLBS4654
Anti-p-ATF2 Rabbit Polyclonal Antibody [clone: S498]
Antikörper
Synthetic phosphopeptide derived from human ATF2 around the phosphorylation site of Serine 498.
p-ATF2 (S498) pAb detects endogenous levels of ATF2 protein only when phosphorylated at Ser498.
Type: Primary
Antigen: p-ATF2
Clonality: Polyclonal
Clone: S498
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse