Anti-p-CASP8 Rabbit Polyclonal Antibody [clone: Y380]

Lieferant: Bioworld Technology
BS4662
BWRLBS4662EA 284 EUR
BWRLBS4662
Anti-p-CASP8 Rabbit Polyclonal Antibody [clone: Y380]
Antikörper
Synthetic phosphopeptide derived from human Caspase-8 around the phosphorylation site of Tyrosine 380.

p-Caspase 8 (Y380) pAb detects endogenous levels of Caspase 8 protein only when phosphorylated at Tyr380.

Type: Primary
Antigen: p-CASP8
Clonality: Polyclonal
Clone: Y380
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR