Anti-p-CaSR Rabbit Polyclonal Antibody [clone: T888]

Lieferant: Bioworld Technology
BS4663
BWRLBS4663EA 284 EUR
BWRLBS4663
Anti-p-CaSR Rabbit Polyclonal Antibody [clone: T888]
Antikörper
Synthetic phosphopeptide derived from human CaSR around the phosphorylation site of Threonine 888.

p-CaSR (T888) pAb detects endogenous levels of CaSR protein only when phosphorylated at Thr888.

Type: Primary
Antigen: p-CaSR
Clonality: Polyclonal
Clone: T888
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR