Anti-p-Chk1 Rabbit Polyclonal Antibody [clone: S286]

Lieferant: Bioworld Technology
BS4670
BWRLBS4670EA 284 EUR
BWRLBS4670
Anti-p-Chk1 Rabbit Polyclonal Antibody [clone: S286]
Antikörper
Synthetic phosphopeptide derived from human Chk1 around the phosphorylation site of Serine 286.

p-Chk1 (S286) pAb detects endogenous levels of Chk1 protein when phosphorylated at Ser286.

Type: Primary
Antigen: p-Chk1
Clonality: Polyclonal
Clone: S286
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR