Anti-p-FoxO4 Rabbit Polyclonal Antibody [clone: T451]

Lieferant: Bioworld Technology
BS4687
BWRLBS4687EA 284 EUR
BWRLBS4687
Anti-p-FoxO4 Rabbit Polyclonal Antibody [clone: T451]
Antikörper
Synthetic phosphopeptide derived from human FoxO4 around the phosphorylation site of Threonine 451.

p-FoxO4 (T451) pAb detects endogenous levels of FoxO4 protein only when phosphorylated at Thr198.

Type: Primary
Antigen: p-FoxO4
Clonality: Polyclonal
Clone: T451
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR