Anti-p-MEK1/2 Rabbit Polyclonal Antibody [clone: S222/226]
Lieferant: Bioworld Technology
BS4734
BWRLBS4734EA 284 EUR
BWRLBS4734
Anti-p-MEK1/2 Rabbit Polyclonal Antibody [clone: S222/226]
Antikörper
Synthetic phosphopeptide derived from human MEK1 around the phosphorylation site of Serine 222.
p-MEK1/2 (S222/226) pAb detects endogenous levels of MEK-1 protein only when phosphorylated at Ser222. This antibody also detects endogenous levels of MEK-2 protein only when phosphorylated at Thr226.
Type: Primary
Antigen: p-MEK1/2
Clonality: Polyclonal
Clone: S222/226
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat