Anti-p-eIF4E Rabbit Polyclonal Antibody [clone: S209]

Lieferant: Bioworld Technology
BS4748
BWRLBS4748EA 284 EUR
BWRLBS4748
Anti-p-eIF4E Rabbit Polyclonal Antibody [clone: S209]
Antikörper
Synthetic phosphopeptide derived from human eIF4E around the phosphorylation site of Serine 209.

p-eIF4E (S209) pAb detects endogenous levels of eIF4E protein only when phosphorylated at Ser209.

Type: Primary
Antigen: p-eIF4E
Clonality: Polyclonal
Clone: S209
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR