Anti-p-BRCA1 Rabbit Polyclonal Antibody [clone: S1423]

Lieferant: Bioworld Technology
BS4756
BWRLBS4756EA 284 EUR
BWRLBS4756
Anti-p-BRCA1 Rabbit Polyclonal Antibody [clone: S1423]
Antikörper
Synthetic phosphopeptide derived from human BRCA1 around the phosphorylation site of Serine 1423.

p-BRCA1 (S1423) pAb detects endogenous levels of BRCA1 protein only when phosphorylated at Ser1423.

Type: Primary
Antigen: p-BRCA1
Clonality: Polyclonal
Clone: S1423
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR