Anti-p-Synapsin I Rabbit Polyclonal Antibody [clone: S9]
Lieferant: Bioworld Technology
BS4802
BWRLBS4802EA 284 EUR
BWRLBS4802
Anti-p-Synapsin I Rabbit Polyclonal Antibody [clone: S9]
Antikörper
Synthetic phosphopeptide derived from human Synapsin I around the phosphorylation site of Serine 9.
p-Synapsin I (S9) pAb detects endogenous levels of Synapsin I protein when phosphorylated at Ser9.
Type: Primary
Antigen: p-Synapsin I
Clonality: Polyclonal
Clone: S9
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat