Anti-p-PR/PGR Rabbit Polyclonal Antibody [clone: S294]

Lieferant: Bioworld Technology
BS4868
BWRLBS4868EA 284 EUR
BWRLBS4868
Anti-p-PR/PGR Rabbit Polyclonal Antibody [clone: S294]
Antikörper
Synthetic phosphopeptide derived from human PGR around the phosphorylation site of Serine 294.

p-PR/PGR (S190) pAb detects endogenous levels of Progesterone Receptor protein only when phosphorylated at Ser190.

Type: Primary
Antigen: p-PR/PGR
Clonality: Polyclonal
Clone: S294
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR