Anti-p-PDGFR-beta Rabbit Polyclonal Antibody [clone: Y751]
Lieferant: Bioworld Technology
BS5030
BWRLBS5030EA 284 EUR
BWRLBS5030
Anti-p-PDGFR-beta Rabbit Polyclonal Antibody [clone: Y751]
Antikörper
Synthetic phosphopeptide derived from human PDGFR-beta around the phosphorylation site of Tyrosine 751.
p-PDGFR-? (Y751) pAb detects endogenous levels of PDGFR-? protein only when phosphorylated at Tyr751.
Type: Primary
Antigen: p-PDGFR-beta
Clonality: Polyclonal
Clone: Y751
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat