Anti-p-Chk2 Rabbit Polyclonal Antibody [clone: T387]
Lieferant: Bioworld Technology
BS5061
BWRLBS5061EA 284 EUR
BWRLBS5061
Anti-p-Chk2 Rabbit Polyclonal Antibody [clone: T387]
Antikörper
Synthetic phosphopeptide derived from human Chk2 around the phosphorylation site of Threonine 387.
p-CHK2 (T387) pAb detects endogenous levels of CHK2 protein when phosphorylated at Thr387.
Type: Primary
Antigen: p-Chk2
Clonality: Polyclonal
Clone: T387
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat