Anti-p-NMDAzeta1 Rabbit Polyclonal Antibody [clone: S896]
Lieferant: Bioworld Technology
BS5090
BWRLBS5090EA 284 EUR
BWRLBS5090
Anti-p-NMDAzeta1 Rabbit Polyclonal Antibody [clone: S896]
Antikörper
Synthetic phosphopeptide derived from human NMDAzeta1 around the phosphorylation site of Serine 896.
p-NMDAR1 (S896) pAb detects endogenous levels of NMDAR1 protein only when phosphorylated at Ser896.
Type: Primary
Antigen: p-NMDAzeta1
Clonality: Polyclonal
Clone: S896
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat