Anti-eIF4E Rabbit Polyclonal Antibody [clone: R215]
Lieferant: Bioworld Technology
BS5516
BWRLBS5516EA 262 EUR
BWRLBS5516
Anti-eIF4E Rabbit Polyclonal Antibody [clone: R215]
Antikörper
Synthetic peptide, corresponding to amino acids 181-230 of Human eIF4E.
eIF4E (R215) pAb detects endogenous levels of eIF4E protein.
Type: Primary
Antigen: eIF4E
Clonality: Polyclonal
Clone: R215
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat