Anti-HDAC1 Rabbit Polyclonal Antibody [clone: E427]

Lieferant: Bioworld Technology
BS5576
BWRLBS5576EA 262 EUR
BWRLBS5576
Anti-HDAC1 Rabbit Polyclonal Antibody [clone: E427]
Antikörper
Synthetic peptide, corresponding to amino acids 398-446 of Human HDAC1.

HDAC1 (E427) pAb detects endogenous levels of HDAC1 protein.

Type: Primary
Antigen: HDAC1
Clonality: Polyclonal
Clone: E427
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR