Anti-HSD11B1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5588
BWRLBS5588EA 262 EUR
BWRLBS5588
Anti-HSD11B1 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 43-88 of Human HSD11B1.

HSD11B1 pAb detects endogenous levels of HSD11B1 protein.

Type: Primary
Antigen: HSD11B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR