Anti-ADCY9 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5590
BWRLBS5590EA 262 EUR
BWRLBS5590
Anti-ADCY9 Rabbit Polyclonal Antibody
Antikörper
A synthetic peptide corresponding to residues in Human ADCY9.

ADCY9 pAb detects endogenous levels of ADCY9 protein.

Type: Primary
Antigen: ADCY9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR