Anti-ALS Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5607
BWRLBS5607EA 262 EUR
BWRLBS5607
Anti-ALS Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 177-222 of Human ALS.

ALS pAb detects endogenous levels of ALS protein.

Type: Primary
Antigen: ALS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR