Anti-SLC25A12 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5616
BWRLBS5616EA 262 EUR
BWRLBS5616
Anti-SLC25A12 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 435-480 of Human SLC25A12.

SLC25A12 pAb detects endogenous levels of SLC25A12 protein.

Type: Primary
Antigen: SLC25A12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR