Anti-BST2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5634
BWRLBS5634EA 262 EUR
BWRLBS5634
Anti-BST2 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 124-169 of Human BST2.

BST2 pAb detects endogenous levels of BST2 protein.

Type: Primary
Antigen: BST2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR