Anti-CA6 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5637
BWRLBS5637EA 262 EUR
BWRLBS5637
Anti-CA6 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to the 263-308 of Human CA6.

CA6 pAb detects endogenous levels of Carbonic anhydrase 6 protein.

Type: Primary
Antigen: CA6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR