Anti-LY75 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5653
BWRLBS5653EA 262 EUR
BWRLBS5653
Anti-LY75 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 509-554 of Human LY75.

LY75 pAb detects endogenous levels of LY75 protein.

Type: Primary
Antigen: LY75
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR