Anti-CD206 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5654
BWRLBS5654EA 262 EUR
BWRLBS5654
Anti-CD206 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 685-730 of Human CD206.

CD206 pAb detects endogenous levels of CD206 protein.

Type: Primary
Antigen: CD206
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR