Anti-CD32-C Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5656
BWRLBS5656EA 262 EUR
BWRLBS5656
Anti-CD32-C Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 278-323 of Human CD32-C.

CD32-C pAb detects endogenous levels of CD32-C protein.

Type: Primary
Antigen: CD32-C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR