Anti-CD58 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5657
BWRLBS5657EA 262 EUR
BWRLBS5657
Anti-CD58 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 78-123 of Human CD58.

CD58 pAb detects endogenous levels of CD58 protein.

Type: Primary
Antigen: CD58
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR