Anti-Cerebellin 1 Rabbit Polyclonal Antibody
Lieferant: Bioworld Technology
BS5658
BWRLBS5658EA 262 EUR
BWRLBS5658
Anti-Cerebellin 1 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 148-193 of Human Cerebellin 1.
Cerebellin 1 pAb detects endogenous levels of Cerebellin 1 protein.
Type: Primary
Antigen: Cerebellin 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat