Anti-CHP2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5661
BWRLBS5661EA 262 EUR
BWRLBS5661
Anti-CHP2 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 146-191 of Human CHP2.

CHP2 pAb detects endogenous levels of CHP2 protein.

Type: Primary
Antigen: CHP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR