Anti-CHST1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5662
BWRLBS5662EA 262 EUR
BWRLBS5662
Anti-CHST1 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 126-171 of Human CHST1.

CHST1 pAb detects endogenous levels of CHST1 protein.

Type: Primary
Antigen: CHST1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR