Anti-CLCNKA Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5665
BWRLBS5665EA 262 EUR
BWRLBS5665
Anti-CLCNKA Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 626-671 of Human CLCNKA.

CLCNKA pAb detects endogenous levels of CLCNKA protein.

Type: Primary
Antigen: CLCNKA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR