Anti-COMT Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5671
BWRLBS5671EA 262 EUR
BWRLBS5671
Anti-COMT Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 111-156 of Human COMT.

COMT pAb detects endogenous levels of COMT protein.

Type: Primary
Antigen: COMT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR