Anti-CPN2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5676
BWRLBS5676EA 262 EUR
BWRLBS5676
Anti-CPN2 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to the 490-535 of Human CPN2.

CPN2 pAb detects endogenous levels of Carboxypeptidase N regulatory subunit protein.

Type: Primary
Antigen: CPN2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR