Anti-CYP11A1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5680
BWRLBS5680EA 262 EUR
BWRLBS5680
Anti-CYP11A1 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 457-502 of Human CYP11A1.

CYP11A1 pAb detects endogenous levels of CYP11A1 protein.

Type: Primary
Antigen: CYP11A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR