Anti-CYP2E1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5686
BWRLBS5686EA 262 EUR
BWRLBS5686
Anti-CYP2E1 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to the 411-456 of Human CYP2E1.

CYP2E1 pAb detects endogenous levels of CYP2E1 protein.

Type: Primary
Antigen: CYP2E1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR