Anti-ELMO1 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5711
BWRLBS5711EA 262 EUR
BWRLBS5711
Anti-ELMO1 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to the 81-126 of Human ELMO1.

ELMO1 pAb detects endogenous levels of ELMO1 protein.

Type: Primary
Antigen: ELMO1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR