Anti-CFI Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5716
BWRLBS5716EA 262 EUR
BWRLBS5716
Anti-CFI Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 492-537 of Human CFI.

CFI pAb detects endogenous levels of CFI protein.

Type: Primary
Antigen: CFI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR