Anti-ZNF801 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5792
BWRLBS5792EA 262 EUR
BWRLBS5792
Anti-ZNF801 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 201-246 of Human ZNF801.

ZNF801 pAb detects endogenous levels of ZNF801 protein.

Type: Primary
Antigen: ZNF801
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR