Anti-MPP9 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5800
BWRLBS5800EA 262 EUR
BWRLBS5800
Anti-MPP9 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 81-126 of Human MPP9.

MPP9 pAb detects endogenous levels of MPP9 protein.

Type: Primary
Antigen: MPP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR