Anti-MTERF Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5804
BWRLBS5804EA 262 EUR
BWRLBS5804
Anti-MTERF Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 311-356 of Human MTERF.

MTERF pAb detects endogenous levels of MTERF protein.

Type: Primary
Antigen: MTERF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR