Anti-MX2 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5805
BWRLBS5805EA 262 EUR
BWRLBS5805
Anti-MX2 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 414-459 of Human MX2.

MX2 pAb detects endogenous levels of MX2 protein.

Type: Primary
Antigen: MX2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR