Anti-Na+ CP type IXalpha Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5813
BWRLBS5813EA 262 EUR
BWRLBS5813
Anti-Na+ CP type IXalpha Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 702-747 of Human Na+ CP type IXalpha.

Na+ CP type IX? pAb detects endogenous levels of Na+ CP type IX? protein.

Type: Primary
Antigen: Na+ CP type IXalpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR