Anti-PGLS Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
BS5852
BWRLBS5852EA 262 EUR
BWRLBS5852
Anti-PGLS Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids 213-258 of Human PGLS.

PGLS pAb detects endogenous levels of PGLS protein.

Type: Primary
Antigen: PGLS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR